VWR®, FAME test method for diesel and petro/biodiesel (6 comp, in n-heptane), CRM
91681.050
:
- Pack type:Glass ampoule/vial
- Pk:5 mL
- Standards:CRM ISO 17034
- Description:FAME Test method for diesel and petro/biodiesel (6 comp, in n-heptane)
- Environmentally Preferable:
- UN No.:1206
- Storage temperature:Ambient (>5 °C)/Sonicate
At stated conc. (µg/ml): Palmitic acid methyl ester (800); Stearic acid methyl ester (800); Methyl cis-9-octadecenoate (800); Linoleic acid methyl ester (800); Chrysene (400); Linolenic acid methyl ester (800).
: Certified reference material.
: Highly Flammable.