Extractables and Leachables Screening Standard for GC, certified reference material, TraceCERT®, 14 components in tert-Butyl methyl ether, Supelco®
Catalog # SUPL01829-1ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass ampoules
- Pk:1 mL
- Description:Extractables and Leachables Screening Standard for GC
- Environmentally Preferable:
- M-Clarity quality level:MQ300
- Storage temperature:−20°C
- Type:Multi-component solution
Specifications
About this item
Ready to use certified reference material mixture suitable for dentification and quantification of extractables and Leachables by GC. 14 components in a concentration of 50 μg/mL.
- Certification of each individual component by qNMR (following ISO 17025 accreditation)
- Mixes produced following the ISO 17034 workflow
- Tested for homogeneity and long-term stability using GC-MS
- Traceability to NIST SRM
- Supplied with a comprehensive certificate including the overall uncertainty
Components:
Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate
ε-Caprolactam
2-Mercaptobenzothiazole
Bisphenol A
3,5-Di-tert-4-butylhydroxytoluene (BHT)
1,3-Di-tert-butylbenzene
Oleamide
Bis(2-ethylhexyl) phthalate
Stearic acid
cis-13-Docosenoamide
Tris(2,4-di-tert-butylphenyl)phosphate
2,4-Di-tert-butylphenol
2,6-Di-tert-butylphenol
Palmitic acid