1,5-Naphthyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:254-79-5
:
Formula:C₈H₆N₂
MDL Number:MFCD00039727
Boiling Pt:150…152 °C/54mmHg
Molecular Weight:130.15 g/mol
Melting Pt:69…75 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together