Ethyl-5-hydroxy-1-methyl-1H-pyrazole-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Ethyl 5-hydroxy-1-methyl-1H-pyrazole-3-carboxylate
CAS Number:51986-17-5
:
Formula:C₇H₁₀N₂O₃
MDL Number:MFCD09839084
Molecular Weight:170.17 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together