3-Bromo-2,2-bis(bromomethyl)propanol
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-Bromo-2,2-bis(bromomethyl)-1-propanol, Tribromoneopentyl alcohol, Pentaerythritol tribromide, 2,2,2-Tris(bromomethyl)ethanol
CAS Number:1522-92-5
:
Formula:C5H9Br3O
MDL Number:MFCD00021982
Boiling Pt:131 °C/2.5mmHg
Molecular Weight:324.8364 g/mol
Melting Pt:66…69 °C
:GHS07
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together