Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acid Red 183
Acid Red 183 30% (dye content), Sigma-Aldrich®
Acid Red 183 30% (dye content), Sigma-Aldrich®
201820-25G
:
SIGMA ALDRICH MICROSCOPY
:
Acid Red 183 30% (dye content), Sigma-Aldrich®
201820-25G
:
SIGMA ALDRICH MICROSCOPY
:
Pk:
25 g
Seq:
3308
Environmentally Preferable:
:
:
532.84 g/mol
:
Ambient
:
6408-31-7
:
C₁₆H₁₁ClN₄Na₂O₈S₂