Order Entry
Austria
ContactUsLinkComponent
1-Naphthyl acetate solution, Sigma-Aldrich®
1-Naphthyl acetate solution, Sigma-Aldrich®
Catalog # 916-10ML
CAS Number:  
1-Naphthyl acetate solution, Sigma-Aldrich®
Catalog # 916-10ML
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Naphthalen-1-yl acetate, Ethanoic acid 1-naphthyl ester, Acetic acid 1-naphthyl ester, α-Naphthyl acetate, Acetic acid α-naphthyl ester, Ethanoic acid α-naphthyl ester
 :
 :186.21 g/mol
 :GHS02
 :GHS06
 :GHS08
Storage Temperature:Refrigerator
 :1
CAS Number:830-81-9
Formula:C₁₂H₁₀O₂
MDL Number:MFCD00003922
Boiling Pt:310
ADR:3 III