Examination gloves, nitrile, Microflex® LifeStar™ EC 93-868
: Ansell
These nitrile gloves provide two layers of reliable barrier protection against hazardous substances and high-risk patient handling.
- Dual layer, dual color design for two layers of protection
- Tested for protection from hazardous chemicals
- Advanced barrier protection (AQL 0.65)
- Non-stick and non-foaming properties for reduced interference and increased productivity
Recommended for: Administering drugs; inspection, selecting, checking parts; sample taking and processing; transferring liquids and solids
: EN 455, EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS; EN 421; Cat. III; AQL 0,65
Food contact approved
Food contact approved
: 100 gloves/dispenser. 90 gloves/dispenser for size 11,5 - 12.
- Catalog No:
- ANSE93-868/8.5-9
- ANSE93-868/9.5-10
- ANSE93-868/10.5-11
- ANSE93-868/6.5-7
- ANSE93-868/7.5-8
- ANSE93-868/11.5-12
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Cuff style:
- Extended
- Extended
- Extended
- Extended
- Extended
- Extended
- Finger thickness:
- 0,21 mm
- 0,21 mm
- 0,21 mm
- 0,21 mm
- 0,21 mm
- 0,21 mm
- Palm thickness:
- 0,14 mm
- 0,14 mm
- 0,14 mm
- 0,14 mm
- 0,14 mm
- 0,14 mm
- EN norm:
- EN 455, EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS; EN 421
- EN 455, EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS; EN 421
- EN 455, EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS; EN 421
- EN 455, EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS; EN 421
- EN 455, EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS; EN 421
- EN 455, EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS; EN 421
- Food contact approved:
- +
- +
- +
- +
- +
- +
Frequently Bought Together