Order Entry
Austria
ContactUsLinkComponent
Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
Certificates
undefined
Phosphotungstic acid hydrate
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to yellow to beige to grey Powder or crystals
  • Assay:
    ≥82 % (WO3)
  • Chloride (Cl):
    ≤20 ppm
  • Sulfate (SO4):
    ≤50 ppm
  • Ammonium (NH4):
    ≤50 ppm
  • Iron (Fe):
    ≤30 ppm
  • Sodium (Na):
    ≤100 ppm
  • Heavy metals (as Pb):
    ≤40 ppm
  • Loss on ignition:
    ≤17 % (800°C)

Specification Test Results

Related Information

Frequently Bought Together