Ethyl-beta-D-glucuronide, certified reference material, Cerilliant®, Supelco®
E-015-1ML
: Merck
:
- Concentration:1,0 mg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:17685-04-0
- Description:Ethyl-beta-D-glucuronide
- Environmentally Preferable:
- Formula:C₈H₁₄O₇
- Description:Ethyl-beta-D-glucuronide
- Pk Info:Ampoule of 1 ml
Ethyl ß-D-glucuronide is a primary urinary metabolite of alcohol and can be used as a biomarker for the monitoring of alchohol consumption.
This Snap-N-Spike® Reference Solution is applicable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing.
: Snap-N-Spike®/Snap-N-Shoot®.