Glucose standard for IC, TraceCERT®, Supelco®
Catalog # 69222-50ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/l
- Pk:50 mL
- Description:Glucose in water
- Environmentally Preferable:
Specifications
About this item
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.