Order Entry
Austria
ContactUsLinkComponent
2-Pyrrolidinone hydrotribromide 97%, Sigma-Aldrich®
2-Pyrrolidinone hydrotribromide 97%, Sigma-Aldrich®
Supplier:  Merck
Certificates
2-Pyrrolidinone hydrotribromide 97%, Sigma-Aldrich®
Supplier:  Merck

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    97%
  • Infrared Spectrum:
    Conforms to Structure
  • Titration by Na₂S₂O₃ :
    96.5 - 103.5%

Specification Test Results

Frequently Bought Together