Rhodium(III) chloride Rh 48,6-49,2%, anhydrous
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Rhodium(III) chloride, Rhodium trichloride
CAS Number:10049-07-7
:
Formula:Cl₃Rh
MDL Number:MFCD00011204
Boiling Pt:717 °C (730 mmHg)
Melting Pt:450 °C
Density:5.38 g/cm³
Molecular Weight:209.26 g/mol
Storage Temperature:Ambient
EINECS:233-165-4
ADR:8 III
Merck Index:12,08355
UN:3260
:GHS05
:GHS07
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Red, grey, red-black or black Crystalline powder, crystals or granules
- Assay:48.6 to 49.2 % (Rh)
- Total trace metal impurities:≤0.001 %
Specification Test Results
Frequently Bought Together