(3-(Methylamino)propyl)trimethoxysilane 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (3-Methylaminopropyl)trimethoxysilane, Trimethoxy[3-(methylamino)propyl]silane, N-Methyl-3-(trimethoxysilyl)propylamine, Trimethoxy(3-(methylamino)propyl)silane
CAS Number:3069-25-8
:
Formula:H₃CNHCH₂CH₂CH₂Si(OCH₃)₃
MDL Number:MFCD00048021
Boiling Pt:106 °C (30 mmHg)
Density:0.978
Molecular Weight:193.32 g/mol
Flash Pt:82 °C (179 °F)
Storage Temperature:Ambient
EINECS:221-334-5
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together