Order Entry
Austria
Orders LinkContactUsLinkComponent
Lead(II) chromate

Lead(II) chromate

Plastic bottle for solids, 880 g
Catalog # VWRS30381.288
Health
Environment
Lead(II) chromate
Catalog # VWRS30381.288
Supplier:  
CAS Number:  7758-97-6
Lead(II) chromate

Plastic bottle for solids, 880 g

CAS Number:  7758-97-6
Discontinued
Product Details & Documents
Synonyms:  Lead chromate
  • CAS Number:7758-97-6
  • Formula:CrO₄Pb
  • MDL Number:MFCD00011158
  • Molecular Weight:323.19 g/mol
  • Merck Index:12,05422
  • Melting Pt:844 °C
  • Density:6.023 g/cm³ (20 °C)
  • EINECS:231-846-0
  • UN:2291
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    880 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.