Potassium hexafluorophosphate 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17084-13-8
:
Formula:F₆KP
MDL Number:MFCD00011412
Molecular Weight:184.06 g/mol
Melting Pt:575
Storage Temperature:Ambient
EINECS:241-143-0
UN:3260
ADR:8 II
:GHS07
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Assay:≥98.5 %
- Loss on drying:≤0.1 % (100°C)
- Free fluoride:≤0.5 %
Specification Test Results
Frequently Bought Together