Salicyl alcohol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (2-Hydroxyphenyl)methanol, 2-(Hydroxymethyl)phenol, 2-Hydroxybenzyl alcohol, Saligenin
CAS Number:90-01-7
:
Formula:C₇H₈O₂
MDL Number:MFCD00004617
Boiling Pt:268
Melting Pt:83…87 °C
Density:1.1613 g/cm³ (25 °C)
Molecular Weight:124.14 g/mol
Merck Index:12,08477
EINECS:201-960-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together