Methyl-3-nitrobenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Methyl 3-Nitrobenzoate, 3-Nitrobenzoic acid methyl ester
CAS Number:618-95-1
:
Formula:C₈H₇NO₄
MDL Number:MFCD00007250
Boiling Pt:279 °C
Molecular Weight:181.15 g/mol
Melting Pt:77…80 °C
Storage Temperature:Ambient
EINECS:210-573-0
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together