Cycloheximide
Catalog # EHERC11830000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
- Environmentally Preferable:
Specifications
About this item
Synonyms: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
:
:281.35 g/mol
EINECS:200-636-0
UN:2811
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:66-81-9
Formula:C₁₅H₂₃NO₄
MDL Number:MFCD00082346
ADR:6.1 I