Taurine ≥99%, white crystalline powder
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: 2-Aminoethanesulphonic acid
CAS Number:107-35-7
:
Formula:C₂H₇NO₃S
MDL Number:MFCD00008197
Molecular Weight:125.15 g/mol
Melting Pt:300
Merck Index:13,09163
Storage Temperature:Ambient
EINECS:203-483-8
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together