Order Entry
Austria
ContactUsLinkComponent
N,N,N',N'-Tetramethylethylenediamine (TEMED) for synthesis, Sigma-Aldrich®
N,N,N',N'-Tetramethylethylenediamine (TEMED) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
N,N,N',N'-Tetramethylethylenediamine (TEMED) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Melting Point:
    -55 °C
  • Molar Mass:
    116.21 g/mol
  • Boiling Point:
    121 °C (1013 hPa)
  • Vapor Pressure:
    21 hPa (20 °C)
  • Flash Point:
    17 °C
  • Explosion Limit:
    1 - 9 %(V)
  • Density:
    0.78 g/cm3 (20 °C)
  • pH:
    8.0 - 8.5 (0.1 g/l, H2O, 20 °C)
  • Synonyms:
    TEMED, TMEDA, 1,2-Bis(dimethylamino)ethane
  • Hill Formula:
    C6H16N2
  • Chemical Formula:
    (CH3)2NCH2CH2N(CH3)2

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together