Order Entry
Austria
ContactUsLinkComponent
2,2-Dichloroacetamide for synthesis, Sigma-Aldrich®
2,2-Dichloroacetamide for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
2,2-Dichloroacetamide for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    71 g/l (20 °C)
  • Melting Point:
    96 - 99 °C
  • Molar Mass:
    127.95 g/mol
  • Boiling Point:
    233 - 234 °C

Specification Test Results

Related Information

Frequently Bought Together