Iron(II) chloride tetrahydrate 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ferrous chloride tetrahydrate, Iron dichloride tetrahydrate
CAS Number:13478-10-9
:
Formula:FeCl₂·4H₂O
MDL Number:MFCD00149709
Molecular Weight:198.81 g/mol
Melting Pt:105…110 °C
Storage Temperature:Ambient
EINECS:231-843-4
UN:3260
ADR:8 II
:GHS07
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow to green Crystalline powder, crystals and/or chunks
- Titration with KMnO4:≥99.0 %
- Sulfate (SO4):≤200 ppm
- Arsenic (As):≤5 ppm
- Copper (Cu):≤50 ppm
- Lead (Pb):≤50 ppm
Specification Test Results
Frequently Bought Together