Potassium hexacyanoferrate(II) trihydrate 99+% for analysis
: Potassiumhexacyanoferrate(II) trihydrate, Tetrapotassium hexacyanoferrate trihydrate, Potassium ferrocyanide trihydrate
:14459-95-1
:
:K₄Fe(CN)₆·3H₂O
:MFCD00167023
:1.85 g/cm³
:422.39 g/mol
:70 °C
:12,07793
:237-722-2
:GHS09
:Ambient
- Environmentally Preferable:
- Appearance:Yellow Crystalline powder
- Titration with KMnO4:≥99.0 %
- Chloride (Cl):≤100 ppm
- Sulfate (SO4):≤100 ppm
- Insoluble matter:≤0.005 %
Frequently Bought Together