Polymyxin B sulphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1405-20-5
:
Formula:C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
MDL Number:MFCD00131991
Molecular Weight:1385.61 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together