Order Entry
Austria
ContactUsLinkComponent
Allyl bromide 99% stabilised
Allyl bromide 99% stabilised
Certificates
undefined
Allyl bromide 99% stabilised
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4680 to 1.4700 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Stabilizer:
    300 ppm propylene oxide

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together