Tantalum ≥99.9% (metals basis), pellets, Ø 1 mm (0.04 in) x 2 mm (0.08 in)
Catalog # 44367.KO
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1.000
- Environmentally Preferable:
Specifications
About this item
CAS Number:7440-25-7
:
Formula:Ta
MDL Number:MFCD00011252
:180.95 g/mol
Boiling Pt:5427
Melting Pt:2996
Density:16.69 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-135-5
Merck Index:12,09223