Glassy carbon, plate, Thickness: 4 mm (0.16 in) Type 2
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-44-0
:
Formula:C
MDL Number:MFCD00133992
Density:1.95 g/cm³ (20 °C)
Molecular Weight:12.01 g/mol
Melting Pt:3652
Merck Index:13,01818
Storage Temperature:Ambient
EINECS:231-153-3
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together