ContactUsLinkComponent
Accessories for goggle gear 500
Accessories for goggle gear 500
Catalog # 111-2371
Supplier:  3M
undefined
Accessories for goggle gear 500
Catalog # 111-2371
Supplier:  3M
Specifications
  • Description:
    3M™ Goggle Gear™ transparent plastic prescription insert for Gear™ 500 GAF
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
3M™ Goggle Gear™ transparent plastic prescription insert can be easily placed in the 3M™ Goggle Gear™ 500 (MMMAGG501SGAF-EU).
  • For polycarbonate prescription lenses up to + and - 6 dioptre
  • This lens is helpful for anyone who needs corrective vision to go along with eye protection
  • Insert available for 3M™ Goggle Gear, 500-Series Splash Goggles