Human enterostatin
ENZOBMLP2340001
New Product
:
- Pk:1 mg
- Source:
- Species:Human
- Seq:02
- Environmentally Preferable:
- Protein/peptide name:Enterostatin
- Purity:95%
Reduces food intake
This pentapeptide, APGPR, has a number of physiological activities, including inhibition of insulin secretion, suppression of fat intake, and memory enhancement. In vivo, it is generated by the cleavage of its precursor protein, procolipase.