Anti-RNF43 Polyclonal Antibody
Catalog # BSBTA01694-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:RNF43
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Rat,Mouse
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for RNF43 detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: RNF43
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse; Rat