Hydroxypropyl methylcellulose (Hypromellose), 50 mPa.s (2% in H₂O) USP Substitution Type 2910
Catalog # SPCMHY122-125GM
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pk:125 g
- Seq:3445
- Environmentally Preferable:
Specifications
Specification Test Results
- Viscosity of 2% aqueous solution@ 20 °C:Min. 40 - 60 mPa.s
- Assay (methoxy groups):Min. 28,0 - 30,0%
- pH:Min. 5,0 - 8,0
- Losss on drying:Max. 5,0%
- Residue on ignition:Max. 1,5%
- Heavy metals:20 ppm
- Identification:To pass test
- Residual solvents:To pass test
Specification Test Results
About this item
Synonyms: Methocel K100, Hydroxypropyl methyl cellulose
:
CAS Number:9004-65-3
Formula:C₃H₈O₂·xCH₄O
MDL Number:MFCD00131360