Order Entry
Austria
ContactUsLinkComponent
Human recombinant TGF beta 3 (from cells)
Human recombinant TGF beta 3 (from cells)
Catalog # PRSI92-444
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant TGF beta 3 (from cells)
Catalog # PRSI92-444
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Environmentally Preferable:
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Transforming growth factor beta-3|TGF-beta-3|Latency-associated Peptide|TGFB3|LAP
  • Protein/peptide name:
    TGF beta 3
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    12.7 kD
  • Sequence:
    Ala301-Ser412(Tyr340Phe
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 4 mM HCl. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Transforming growth factor beta 3(TGFB3) is a member of a TGF - beta superfamily which is defined by theirstructural and functional similarities. TGFB3 is secreted as a complex with LAP. This latent form of TGFB3becomes active upon cleavage by plasmin, matrix metalloproteases, thrombospondin -1, and a subset ofintegrins. It binds with high affinity to TGF- beta RII, a type II serine/threonine kinase receptor. TGFB3 is involved incell differentiation, embryogenesis and development.It is believed to regulate molecules involved in cellularadhesion and extracellular matrix (ECM) formation during the process of palate development. Without TGF- beta 3,mammals develop a deformity known as a cleft palate.

This recombinant protein can be used for biological assays. For research use only.