Order Entry
Austria
ContactUsLinkComponent
 
Strep-tag II peptide
  PRSI4217P
 :  
undefined
Strep-tag II peptide
  PRSI4217P
 :  

 

  • Pk:
    50 µG
  • Source:
  • Species:
  • Environmentally Preferable:
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Protein/peptide name:
    Strep-tag II Peptide
  • Tested applications:
    Blocking

 

 

Strep-tag II peptide is used for blocking the activity of strep-tag II antibody.