Specifications
- Antigen name:Solute Carrier Family 6 (neurotransmitter transporter GABA) Member 1
- Antigen symbol:SLC6A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:GABT1|GABAT1|GABATR|Solute carrier family 6 member 1|GAT-1|GABA T-1|GABA T1|GAT1|Sodium- and chloride-dependent GABA transporter 1
- Immunogen:Peptide corresponding to amino acid residues from the C-terminal region of rat GAT-1.
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Gamma-aminobutyric acid (GABA) is the primary inhibitory neurotransmitter in the central nervous system, causing a hyperpolarization of the membrane through the opening of a Cl− channel associated with the GABAA receptor (GABAA-R) subtype. GABA plasma membrane transporters (GATs) influence synaptic neurotransmission by high-affinity uptake and release of GABA. To date, four distinct GABA transporters have been identified: GAT-1, GAT-2, GAT-3, and BGT-1. GAT-1, the most abundant of the transporters, is found predominantly in neurons, but also in some specialized glia (Minelli et al., 1995). GAT-1 is thought to play a key role in epileptogenesis (Zhao et al. 2003).
The antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that the antibody will also react with mouse tissue based on the fact that this species has 100% homology with the amino acid sequence used as antigen.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Slc6a1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat