Anti-ENG Rabbit Polyclonal Antibody
Catalog # PRSI27-781
Supplier: ProSci Inc.
Specifications
- Antigen name:Endoglin
- Antigen symbol:ENG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:ENG|END|ORW1|HHT1
- Immunogen:A synthetic peptide corresponding a region of human ENG.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Endoglin is a homodimeric transmembrane glycoprotein highly expressed by endothelial cells. It is a component of the transforming growth factor beta receptor complex since it binds TGFB1 and TGFB3 with high affinity. Mutations in the endoglin gene produce hereditary hemorrhagic telangiectasia.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ENG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human