Specifications
- Antigen name:Acyl-CoA Dehydrogenase, C-4 to C-12 straight chain
- Antigen symbol:ACADM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:ACAD1|C-4 to C-12 straight chain|FLJ18227|ACADM|medium-chain specific acyl-CoA dehydrogenase|acyl-Coenzyme A dehydrogenase|RP4-682C21.1|medium-chain acyl-CoA dehydrogenase|MCADH|MCAD|FLJ99884|FLJ93013
- Immunogen:A synthetic peptide corresponding a region of human ACADM.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
ACADM Is the medium-chain specific (C4 to C12 straight chain) acyl-Coenzyme A dehydrogenase. The homotetramer enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Clinical phenotypes are associated with ACADM hereditary deficiency.This gene encodes the medium-chain specific (C4 to C12 straight chain) acyl-Coenzyme A dehydrogenase. The homotetramer enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Defects in this gene cause medium-chain acyl-CoA dehydrogenase deficiency, a disease characterized by hepatic dysfunction, fasting hypoglycemia, and encephalopathy, which can result in infantile death. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
ACADM antibody can be used for detection of ACADM by ELISA at 1:1562500. ACADM antibody can be used for detection of ACADM by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ACADM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human