Order Entry
Austria
ContactUsLinkComponent
 
Anti-4E-BP1 Rabbit Polyclonal Antibody
  PRSI3513
undefined
Anti-4E-BP1 Rabbit Polyclonal Antibody
  PRSI3513

 

  • Antigen name:
    Eukaryotic translation initiation factor 4E-binding protein 1
  • Antigen symbol:
    4E-BP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Antigen synonyms:
    PHAS-1|PHAS-I|Insulin-stimulated EIF-4E binding protein PHAS-I|P/OKCL.6|EIF4EBP1|4EBP1
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    4E-BP1 antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human 4E-BP1.
  • Tested applications:
    E, WB, ICC
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

4E-BP1 Antibody: The translation of mRNA in eukaryotic cells is regulated by the presence of amino acids through multiple mechanisms. One such mechanism involves the evolutionarily conserved serine/threonine kinase TOR (Target of rapamycin, also known as mTOR), which regulates cell growth and cell cycle through its ability to integrate signals from nutrient levels and growth factors. One downstream target of TOR is the eukaryotic initiation factor 4E binding protein 1 (4E-BP1) whose phosphorylation prevents its association with eIF4E, preferentially stimulating translation of mRNAs containing long, highly structured 5'-UTRs. Rapamycin inhibits TOR resulting in reduced cell growth and reduced rates of cell cycle and cell proliferationl, at least in part by inhibiting the activity of TOR towards 4E-BP1.

Type:
Antigen: 4E-BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse