ContactUsLinkComponent
Peanut butter, NIST-2387 standard
Peanut butter, NIST-2387 standard
  PROONIST-2387
 :  
Peanut butter, NIST-2387 standard
  PROONIST-2387
 :  
  • Pk:
    3
  • Description:
    NIST-2387 standard, peanut butter
  • Environmentally Preferable:
This reference material intended to use in validating methods for determining fat, fatty acids, trace elements and tocopherol.
 : Traceable to NIST and SRM.