Heated product shelf chambers for FreeZone® freeze dryer
Supplier: LABCONCO
Certificates
About this item
Heated product shelves with clear chamber for drying bulk samples, or drying serum bottles, vials or microplates. Chamber connects to a FreeZone® freeze dryers for control and recording of the shelf temperature from the Lyo-Works display.
- Wth removable 254 mm Ø shelves
- Each shelf is height adjustable to accommodate a broad variety of sample sizes
- Shelf temperature may be set from –20 to 60 °C
- Shelves may be placed in freezer for pre-freezing
Each chamber includes a 305 mm Ø clear acrylic cylinder, vacuum release valve and 203 mm Ø opening to collector at the bottom of chamber. Can be fitted with 257 or 356 mm Ø condensers.
Short chambers include three shelves and three temperature probes. Tall chambers include five shelves and five temperature probes.
Certifications: ETL listed and CE marked.
Ordering information: FreeZone® freeze dryer with Lyo-Works™ OS required (not included).
Frequently Bought Together