Cold resistant gloves, ActivArmr® 23-700
: Ansell
These fully coated gloves feature a PU foam insulation for additional warmth and comfort. The textured surface provides unbeatable grip on wet and slippery surfaces. The robust, liquid-repellent PVC coating does not absorb water, does not stiffen like leather and remains flexible even in sub zero temperatures.
- Salt, chemical and corrosion resistant - does not crack or split
- Ergonomic fit for increased comfort - reduces finger and hand fatigue
- Water-repellent, turn-out lining carries sweat away from hand
Ideal for food processing and automotive applications.
: EN 388:2016 (3121B), EN 511 (111), EN ISO 374-1 Type B (KLT), EN ISO 374-5; Cat. III
- Catalog No:
- 112-2926
- 112-0270
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Liner material:
- Cotton/nylon/PU
- Cotton/nylon/PU
- Finish:
- Fully PVC coated
- Fully PVC coated
- Cuff style:
- Gauntlet
- Gauntlet
- EN norm:
- EN 388:2016 (3121B), EN 511 (111), EN ISO 374-1 Type B (KLT), EN ISO 374-5
- EN 388:2016 (3121B), EN 511 (111), EN ISO 374-1 Type B (KLT), EN ISO 374-5
- Anti-static:
- +
- +
Frequently Bought Together