Order Entry
Austria
ContactUsLinkComponent
Anti-G6PC3 Rabbit Polyclonal Antibody (Cy5®)
Anti-G6PC3 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-13253R-CY5
Supplier:  Bioss
Anti-G6PC3 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-13253R-CY5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glucose 6 Phosphatase, Catalytic, 3
  • Antigen symbol:
    G6PC3
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    92579
  • Antigen synonyms:
    SCN4|UGRP
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Glucose-6-phosphatase (G6Pase), is a multicomponent enzyme system that hydrolyzes glucose-6-phosphate (G6P) in the final step of gluconeogenesis and gluconeolysis. G6Pase localizes to the endoplasmic reticulum, and while liver, kidney, and intestine are the only tissues that express the first identified isoform, G6Pase-Alpha, a second form, designated G6Pase-Beta, contributes to blood glucose homeostasis in a wider range of tissues. G6Pase-Beta, also known as SCN4, UGRP or G6PC3 (glucose 6 phosphatase, catalytic, 3), is a 346 amino acid endoplasmic reticulum multi-pass membrane protein that is involved in carbohydrate biosynthesis and the gluconeogenesis pathway. Inhibited by vanadate, G6Pase-Beta hydrolyzes GP6 to glucose in the endoplasmic reticulum. Due to its necessary involvement in normal glucose metabolism, G6Pase-Beta may play an integral role in diabetes and glycogen storage diseases (GSDs).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: G6PC3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat