Anti-Strep II Tag Mouse Monoclonal Antibody
Catalog # BWRLMB2017
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Strep II Tag
- Clonality:Monoclonal
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Multiple
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide Strep II Tag.
Strep ?tag mouse monoclonal antibody detects endogenous levels of internal, C-terminal, and N-terminal Strep ?- tagged proteins.
Type: Primary
Antigen: Strep II Tag
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: All