Order Entry
Austria
ContactUsLinkComponent
 
Anti-YAP Rabbit Polyclonal Antibody [clone: Q295]
  BWRLBS2000
Anti-YAP Rabbit Polyclonal Antibody [clone: Q295]
  BWRLBS2000

 

  • Antibody type:
    Primary
  • Antigen symbol:
    YAP
  • Clonality:
    Polyclonal
  • Clone:
    Q295
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 280-330 of Human YAP.

YAP (Q295) pAb detects endogenous levels of YAP protein.

Type: Primary
Antigen: YAP
Clonality: Polyclonal
Clone: Q295
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat