Anti-ADCY 4 Rabbit Polyclonal Antibody [clone: L226]
Catalog # BWRLBS3421
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:ADCY 4
- Clonality:Polyclonal
- Clone:L226
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 190-240 of Human ADCY 4.
ADCY 4 (L226) pAb detects endogenous levels of Adenylate cyclase type 4 protein.
Type: Primary
Antigen: ADCY 4
Clonality: Polyclonal
Clone: L226
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat