McFarland turbidity standards, Remel™
These McFarland equivalence turbidity standard is packed in plastic case with a visual comparision card. Tube size of 15×103 mm.
- Fits in most spectrometers
- Polystyrene technology allows longer shelf life
: The turbidity standard set includes 1 each of 0,5, 1,0, 2,0, 3,0 and 4,0.
- Turbidity:
- Environmentally Preferable:
Frequently Bought Together