Order Entry
Austria
ContactUsLinkComponent
Pig Follicle stimulating hormone (from Pituitary), MP Biomedicals
Pig Follicle stimulating hormone (from Pituitary), MP Biomedicals
Catalog # ICNA0210172750
Supplier:  MP Biomedicals
CAS Number:  
Pig Follicle stimulating hormone (from Pituitary), MP Biomedicals
Catalog # ICNA0210172750
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    50 AU
  • CAS No.:
    9002-68-0
  • Source:
    Pituitary
  • Species:
    Pig
  • Environmentally Preferable:
  • Storage conditions:
    0 °C
  • Form:
    Powder
  • Protein/peptide name:
    Follicle stimulating hormone
  • Colour:
    Off-white

Specifications

About this item

Follicle stimulating hormone is a two-chain glycoprotein gonadotropic hormone that induces maturation of the Graafian follicles of the ovary. It promotes the development of germinal cells in males. It regulates sertoli cells by acting on G-protein-linked cell surface FSH receptors. It activates cytosolic soluble protein tyrosine kinase (CyPTk).

  • Soluble in water (50 mg/ml)

It is used in conjunction with asparagine 56-deglycosylated eLHα acted as an antagonist for cAMP accumulation in HEK-293 cells stably expressing the FSH-R, a G protein-coupled receptor (GPCR) specifically expressed in somatic cells of the gonads and which plays a crucial role in reproduction.