Anti-SR-BI/SR-BII Rabbit Polyclonal Antibody
Rabbit Polyclonal SR-BI/SR-BII Antibody. Tested Applications: Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry. Tested Reactivity: Mouse.
There is 1 scientific publication using this antibody.
Type: Primary
Antigen: SR-BI/SR-BII
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
- Catalog No:
- NOVUNB400-134
- NOVUNB400-134SS
- Antigen symbol:
- SCARB1
- SCARB1
- Antigen name:
- Scavenger receptor class B, member 1
- Scavenger receptor class B, member 1
- Antigen synonyms:
- SR-BI|HDLQTL6|SRB1|CLA-1|CD36L1|CLA1
- SR-BI|HDLQTL6|SRB1|CLA-1|CD36L1|CLA1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 949
- 949
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together