Anti-B3GAT1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: NK-1]
Catalog # BNUM0016-50
Supplier: Biotium
Specifications
- Antibody type:Primary
- Antigen name:Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1
- Antigen symbol:B3GAT1
- Clonality:Monoclonal
- Clone:Clone NK-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM kappa
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:381050
- Antigen synonyms:LEU7|GLCATP|CD57|NK1|GLCUATP|NK-1|beta-1,3-glucuronyltransferase 1|HNK1
- Storage buffer:PBS/0,05% azide
- Concentration:1 mg/mL
- Purification:BSA-free
- Size:50 μL
- Pk:50 µl
Specifications
About this item
Anti-CD57 marks a subset of lymphocytes known as natural killer (NK) cells. Follicular center cell lymphomas often contain many NK cells within the neoplastic follicles. Anti-CD57 also stains neuroendocrine cells and their derived tumors, including carcinoid tumor and medulloblastoma. Anti-CD57 can also be useful in separating type B3 thymoma from thymic carcinoma when combined with a panel that includes antibodies against GLUT1, CD5, and CEA.
Type: Primary
Antigen: B3GAT1
Clonality: Monoclonal
Clone: NK-1
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human