Order Entry
Austria
ContactUsLinkComponent
 
Toluene, Technical Grade
Catalog # ACRO451970050
CAS Number:  
undefined
Toluene, Technical Grade
Catalog # ACRO451970050
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    5 L
  • Seq:
    5190
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Refractive index:
    1.4900 to 1.5030 (20°C, 589 nm)
  • Density:
    0.865 to 0.872 kg/l

Specification Test Results

About this item

Synonyms: Methylbenzene, Phenylmethane, FM approval
 :
 :92.14 g/mol
UN:1294
 :GHS07
 :GHS08
 :GHS02
Melting Pt:–95 °C
Storage Temperature:Ambient
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512
Boiling Pt:110.6
Density:0.866 g/cm³
Flash Pt:4
ADR:3 II
Merck Index:13,09607